0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standards
Environmental Analysis Standards
Anionic Surfactant - WP, (CRM), Supelco®
Anionic Surfactant - WP, (CRM), Supelco®
Anionic Surfactant - WP, (CRM), Supelco®
Catalog # QC1144-20ML
Supplier:
Merck
CAS Number:
Expand image
Anionic Surfactant - WP, (CRM), Supelco®
Catalog # QC1144-20ML
Supplier:
Merck
CAS Number:
Specifications
Pk:
20 mL
Description:
Anionic Surfactant - WP
Type:
Certified reference material
Specifications
About this item
Product Details
Product Details