Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Standards
Environmental Analysis Standards
Regulated SOCS kit - WS, (CRM), Supelco®
Regulated SOCS kit - WS, (CRM), Supelco®
Regulated SOCS kit - WS, (CRM), Supelco®
Catalog # PE1354-1KT
Supplier:
Merck
CAS Number:
Expand image
Regulated SOCS kit - WS, (CRM), Supelco®
Catalog # PE1354-1KT
Supplier:
Merck
CAS Number:
Specifications
Pk:
1 KIT
Description:
Regulated SOCS kit - WS
Specifications
About this item
Product Details
Product Details