Supelco® Organic Reference Standard, TO11/IP-6A Aldehyde/Ketone-DNPH Mix
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338
Specifications
- Storage conditions:2…8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Flash point:2,0 °C - Closed cup
Specifications
Frequently Bought Together