Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, EPA 8270 Semivolatile Internal Standard Mix
Supelco® Organic Reference Standard, EPA 8270 Semivolatile Internal Standard Mix
Catalog # CRM47906
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, EPA 8270 Semivolatile Internal Standard Mix
Catalog # CRM47906
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    High concentration, 4000 µg/ml each component in methylene chloride: benzene (1:1)
  • Pk:
    1 Ampoul
  • Standards:
    Certified reference material
  • Description:
    EPA 8270 Semivolatile internal standard mix
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H304 - H315 - H319 - H336 - H340 - H350 - H372 - H411
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09
Caution: P210 - P273 - P301 + P310 - P303 + P361 + P353 - P305 + P351 + P338 - P331