Order Entry
Czech Republic
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, BTEX/MTBE Mix, HC
  CRM47505
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, BTEX/MTBE Mix, HC
  CRM47505
 :  Merck
 :  

 

  • Concentration:
    2000 μg/ml each component in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material, TraceCERT®
  • Description:
    BTEX/MTBE mix, HC
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H340 - H350 - H370
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Flash point:
    9,7 °C - Closed cup

 

 

  • GHS signal: GHS02, GHS06, GHS08
 : P202 - P210 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311