Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Environmental Analysis Standards
Clean Clay #5, (CRM), Supelco®
Clean Clay #5, (CRM), Supelco®
Clean Clay #5, (CRM), Supelco®
Catalog # CLNSOIL5-100G
Supplier:
Merck
CAS Number:
Expand image
Clean Clay #5, (CRM), Supelco®
Catalog # CLNSOIL5-100G
Supplier:
Merck
CAS Number:
Specifications
Pk:
100 g
Description:
Clean Clay #5
Specifications
About this item
Product Details
Product Details