Order Entry
Czech Republic
ContactUsLinkComponent
 
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) ≥98% (by TLC) fluorescent pH indicator
Catalog # 51015.
Supplier:  Biotium
CAS Number:  
undefined
5(6)-Carboxy-2',7'-dichlorofluorescein (CDCF) ≥98% (by TLC) fluorescent pH indicator
Catalog # 51015.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

About this item

 :
 :445.204 g/mol
Storage Temperature:Ambient
CAS Number:111843-78-8
Formula:C₂₁H₁₀Cl₂O₇
MDL Number:MFCD00037452