Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Succinimidyl ester acridine orange
Succinimidyl ester acridine orange DNA/RNA binding dye
Succinimidyl ester acridine orange DNA/RNA binding dye
Succinimidyl ester acridine orange DNA/RNA binding dye
Catalog # 40074.
Supplier:
Biotium
CAS Number:
Expand image
Succinimidyl ester acridine orange DNA/RNA binding dye
Catalog # 40074.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
557.49 g/mol
Storage Temperature:
Freezer
Formula:
C₂₇H₃₃BrN₄O₄
Product Details