0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Succinimidyl ester ethidium bromide
Succinimidyl ester ethidium bromide DNA/RNA binding dye
Succinimidyl ester ethidium bromide DNA/RNA binding dye
Catalog # 40072.
Supplier:
Biotium
CAS Number:
Expand image
Succinimidyl ester ethidium bromide DNA/RNA binding dye
Catalog # 40072.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
498 g/mol
Storage Temperature:
Freezer
Product Details