Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, Acetone- 2,4-DNPH solution
Supelco® Organic Reference Standard, Acetone- 2,4-DNPH solution
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Acetone- 2,4-DNPH solution
Supplier:  Merck

Specifications

  • CAS No.:
    1567-89-1
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Acetone-2,4-dinitrophenylhydrazone solution
  • Molecular weight:
    238,20
  • Flash point:
    2,0 °C - Closed cup

Specifications

Frequently Bought Together