Supelco® Organic Reference Standard, Acetaldehyde-2,4-DNPH solution
: Merck
- GHS signal: GHS02, GHS07
: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338
- CAS No.:1019-57-4
- Storage conditions:2…8 °C
- Hazard class description:H225 - H302 + H312 + H332 - H319
- Shipping conditions:WICE
- M-Clarity quality level:MQ100
- Synonym(s):Acetaldehyde-2,4-dinitrophenylhydrazone solution
- Molecular weight:224,17
- Flash point:2,0 °C - Closed cup
Frequently Bought Together