Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Drug and metabolite standards, Quetiapine carboxylic acid-D8 solution, (CRM)
Supelco® Drug and metabolite standards, Quetiapine carboxylic acid-D8 solution, (CRM)
Catalog # Q-005-1ML
Supplier:  Merck
CAS Number:  
Supelco® Drug and metabolite standards, Quetiapine carboxylic acid-D8 solution, (CRM)
Catalog # Q-005-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    Quetiapine carboxylic acid-D8 solution
  • Description:
    Quetiapine carboxylic acid-D8 solution
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item