Supelco® Cerilliant® Organic Reference Standard, Lactoferrin (from bovine milk)
Catalog # L-047-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Certified reference material, Cerilliant®
- CAS No.:146897-68-9
- Description:Lactoferrin (from bovine milk)
- Storage conditions:–20 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ300