Supelco® Drug and metabolite standards, Deschloroetizolam solution, (CRM)
D-156-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:40054-73-7
- Description:Deschloroetizolam solution
- Description:Deschloroetizolam solution
- Pk Info:Ampoule of 1 ml