6-(Fluorescein-5(6)-carboxamido) hexanoic acid succinimidyl ester [5(6)-SFX] mixture of isomers ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90044.
Supplier: Biotium
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:586.55 g/mol
Storage Temperature:Freezer
CAS Number:114116-31-8
Formula:C₃₁H₂₆N₂O₁₀