Order Entry
Czech Republic
ContactUsLinkComponent
 
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90034.
Supplier:  Biotium
CAS Number:  
undefined
5-Carboxytetramethylrhodamine succinimidyl ester (5-TAMRA SE) ≥98% (by TLC) amine-reactive fluorescent dye
Catalog # 90034.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :527.52 g/mol
Storage Temperature:Freezer
CAS Number:150810-68-7
Formula:C₂₉H₂₅N₃O₇