5-Carboxytetramethylrhodamine (5-TAMRA) free acid amine-reactive fluorescent dye
Catalog # 90009.
Supplier: Biotium
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:430.45 g/mol
Storage Temperature:Refrigerator
CAS Number:91809-66-4
:MFCD1073638
Formula:C₂₅H₂₂N₂O₅
MDL Number:MFCD00269769