Supelco® Drug and metabolite standards, Tapentadol-beta-D-glucuronide solution, (CRM)
Catalog # T-060-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile: water
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:130037-86-8
- Description:Tapentadol-beta-D-glucuronide solution
- Description:Tapentadol-beta-D-glucuronide solution
- Pk Info:ampoule of 1 ml