Supelco® Drug and metabolite standards, Spice Cannabinoid Mix 3 solution, (CRM)
Catalog # S-047-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml each component in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- Description:Spice Cannabinoid Mix 3 solution
- Pk Info:ampoule of 1 ml