Supelco® Drug and metabolite standards, JWH-073 solution, (Spice Cannabinoid), (CRM)
Catalog # S-027-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in acetonitrile
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:208987-48-8
- Description:JWH-073 solution, (Spice Cannabinoid)
- Description:JWH-073 solution, (Spice Cannabinoid)
- Pk Info:ampoule of 1 ml