Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Drug and metabolite standards, Androstene-3,17-dione-2,3,4-13C3 solution, (CRM)
Supelco® Drug and metabolite standards, Androstene-3,17-dione-2,3,4-13C3 solution, (CRM)
Catalog # A-084-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Drug and metabolite standards, Androstene-3,17-dione-2,3,4-13C3 solution, (CRM)
Catalog # A-084-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    327048-86-2
  • Description:
    Androstene-3,17-dione-2,3,4-13C3 solution
  • Description:
    Androstene-3,17-dione-2,3,4-13C3 solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item