Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Cerilliant® Organic Reference Standard, Aldosterone solution
Supelco® Cerilliant® Organic Reference Standard, Aldosterone solution
Catalog # A-096-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Cerilliant® Organic Reference Standard, Aldosterone solution
Catalog # A-096-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Certified reference material, Cerilliant®
  • CAS No.:
    52-39-1
  • Description:
    Aldosterone solution
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00051136
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Aldosterone; 11β,21-Dihydroxy-3,20-dioxo-4-pregnen-18-al; 11β,21-Dihydroxypregn-4-ene-3,18,20-trione; 18-Aldocorticosterone; Reichstein X
  • Molecular weight:
    360,44
  • Flash point:
    2 °C - Closed cup
  • Pk Info:
    Ampule of 1 ml

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338