Order Entry
Czech Republic
ContactUsLinkComponent
Drug and metabolite standards, 17alpha-Ethynylestradiol solution, (CRM), Supelco®
Drug and metabolite standards, 17alpha-Ethynylestradiol solution, (CRM), Supelco®
Catalog # E-076-1ML
Supplier:  Merck
CAS Number:  
undefined
Drug and metabolite standards, 17alpha-Ethynylestradiol solution, (CRM), Supelco®
Catalog # E-076-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1,0 mg/ml in methanol
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    57-63-6
  • Description:
    17alpha-Ethynylestradiol solution
  • Description:
    17alpha-Ethynylestradiol solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item