Order Entry
Czech Republic
ContactUsLinkComponent
 
Drug and metabolite standards, (+/-)-MBDB hydrochloride, (CRM), Supelco®
  M-102-1ML
 :  Merck
 :  
undefined
Drug and metabolite standards, (+/-)-MBDB hydrochloride, (CRM), Supelco®
  M-102-1ML
 :  Merck
 :  

 

  • Concentration:
    1,0 mg/ml in methanol (as free base)
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • CAS No.:
    103818-37-7
  • Description:
    (+/-)-MBDB hydrochloride
  • Description:
    (+/-)-MBDB hydrochloride
  • Pk Info:
    ampoule of 1 ml