Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, Linoleic Acid Methyl Ester Mix
Supelco® Organic Reference Standard, Linoleic Acid Methyl Ester Mix
Catalog # CRM47791
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Linoleic Acid Methyl Ester Mix
Catalog # CRM47791
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    cis/trans, 10 mg/ml in dichloromethane
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    Linoleic Acid methyl ester mix
  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H315 - H319 - H336 - H351
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS08, GHS07
Caution: P201 - P202 - P261 - P302 + P352 - P305 + P351 + P338 - P308 + P313