Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, F.A.M.E. Mix GLC-10
Supelco® Organic Reference Standard, F.A.M.E. Mix GLC-10
Catalog # CRM1891
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, F.A.M.E. Mix GLC-10
Catalog # CRM1891
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    Pk of 100 mg (Neat)
  • Pk:
    100 mg
  • Standards:
    Certified reference material
  • Description:
    F.A.M.E. mix GLC-10
  • Storage conditions:
    –10...–25 °C
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100

Specifications

About this item

This certified reference material is useful for determining relative retention times and approximating response factors.