Order Entry
Czech Republic
ContactUsLinkComponent
Middlebrook ADC growth supplement
Middlebrook ADC growth supplement
Catalog # SIALM0553-1VL
Supplier:  Merck
CAS Number:  
Middlebrook ADC growth supplement
Catalog # SIALM0553-1VL
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Middlebrook ADC growth supplement, 1 vial

Specifications

About this item

These supplements are ideal for cultivation of Mycobacteria species.

Reconstitution: Warm up the refrigerated supplement to 45 to 50 °C. Aseptically add the contents of the vial to 450 ml of sterile Middlebrook 7H9 broth base. Mix well and pour into sterile screw capped tubes.

Store at 2 to 8 °C.

Delivery: Components (per vial) include bovine albumin fraction V 2,5 g, dextrose 1,0 g, catalase 0,0015 g and distilled water 50 ml.