Penicillin : Streptomycin solution in 0.9% NaCl, Hybri-Max™, Bioreagent, sterile filtered
Catalog # SIALP7539-100ML
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance (Form):Solution
- Appearance (Turbidity):Clear
- Osmolality:306 - 374 mOs/kg
- Potency - Penicillin:10000 - 12000 U/ml
- Potency Streptomycin:10 - 12 mg/ml
- Sterility:Pass
- Endotoxin Level at 10 ml/l:≤0.5 EU/ml
- Cell Line Suspended Cell Line:Sp2 Mouse Myeloma
- % Cloning Performance:≥80%
Specification Test Results
About this item
:
Storage Temperature:Freezer
CAS Number:1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)
:113-98-4