Iron(II,III) oxide solution 5 mg/ml in toluene, nanoparticles 9 - 11 nm, magnetic, Sigma-Aldrich®
Catalog # SIAL700312-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance (Colour):Dark Brown to Black
- Appearance (Form):Liquid
- ICP Major Analysis, Confirms Iron Component:Confirmed
- Miscellaneous Assay:Concentration:5 mg/ml in toluene
- Magnetisation:≥45 emu/g, at ambient temperature under 4500 Oe
- Particle Size:10 nm +/-1 nm (TEM)
Specification Test Results
About this item
:
:231.54 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:7 °C
UN:1294
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:1317-61-9
Formula:Fe₃O₄
MDL Number:MFCD00011010