

Sulphuric acid fuming 65% (SO₃), GPR RECTAPUR®
Catalog # 86153.243
Supplier: VWR Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, PVC-coated
- Pk:2 Bottle
- Seq:5041
- Pk Info:2× 1 L
Specifications
Specification Test Results
- Content of free SO₃ (acidimetric):64 – 66%
Specification Test Results
About this item
:
Boiling Pt:64 °C (1013 hPa)
Melting Pt:1 °C
Density:1.99 g/cm3 (20 °C)
ADR:8 (6.1),I
:80.06
EINECS:231-976-8
UN:1831
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:8014-95-7
Formula:H₂O₇S₂ (H₂SO₄ + SO₃)(1:2)
MDL Number:MFCD00137792