Order Entry
Czech Republic
ContactUsLinkComponent
Silicic acid hydrate (≥84% SiO₂), powder, BAKER ANALYZED®, J.T.Baker®
Silicic acid hydrate (≥84% SiO₂), powder, BAKER ANALYZED®, J.T.Baker®
Catalog # 0324-01
Supplier:  Avantor
CAS Number:  
undefined
Silicic acid hydrate (≥84% SiO₂), powder, BAKER ANALYZED®, J.T.Baker®
Catalog # 0324-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap, 1000cc
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥84%
  • Assay (as SiO₂):
    ≥84 %
  • Nonvolatile with HF:
    ≤0.4 %
  • Chloride (Cl):
    ≤0.01 %
  • Sulfate (SO₄):
    ≤0.02 %
  • Heavy Metals (as Pb):
    ≤0.002 %
  • Iron (Fe):
    ≤0.007 %
  • Loss on Ignition (as H₂O):
    ≤16 %

Specification Test Results

About this item

 :
 :60.09 g/mol (anhy)
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
Density:2.10 g/cm³
Storage Temperature:Ambient
REACH:01-2119379499-16
CAS Number:63231-67-4
Formula:SiO₂·xH₂O
MDL Number:MFCD00054122