
Methylamine hydroiodide ≥98%, Sigma-Aldrich®
SIAL793493-5G
: Merck
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance (Colour):White to Yellow
- Appearance (Form):Powder or Crystals
- Proton NMR Spectrum:Conforms to Structure
- Titration with AgNO₃:97.5 - 102.5%
- Assay:98%
:
:158.97 g/mol
:270...280 °C
:GHS07
:14965-49-2
:CH₃NH₂·HI
:MFCD28100833