ContactUsLinkComponent
SBW glass stoppers, standard ground joint, octagonal
SBW glass stoppers, standard ground joint, octagonal
Katalogové číslo 217-9125
Dodavatel:  DWK Life Sciences
undefined
SBW glass stoppers, standard ground joint, octagonal
Katalogové číslo 217-9125
Dodavatel:  DWK Life Sciences
Specifikace
  • NS:
    14/23
  • Type:
    Clear glass
  • Seq:
    12
  • Pk:
    1
Specifikace
O tomto produktu
These stoppers are made from SBW glass and have an octagonal head for good grip.
  • Solid form
  • Available amber or clear
Certifications: DIN 12252