Order Entry
Czech Republic
ContactUsLinkComponent
 
Paraffin, Solidification point: 56 - 58 °C, pellets for clinical diagnosis
Catalog # APPC253211.0914
Supplier:  PanReac AppliChem
CAS Number:  
Paraffin, Solidification point: 56 - 58 °C, pellets for clinical diagnosis
Catalog # APPC253211.0914
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Identity:
    IR Passes Test
  • Acidity or Alkalinity:
    Passes Test
  • Insoluble Matter in C₈H₁₀ :
    Passes Test
  • Melting range:
    56 - 58 °C

Specification Test Results

About this item

 :
Boiling Pt:350 °C
Melting Pt:56...58 °C
EINECS:232-315-6
CAS Number:8002-74-2
 :8042-47-5