Order Entry
Czech Republic
ContactUsLinkComponent
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine 97%
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine 97%
Supplier:  Merck
Certificates
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine 97%
Supplier:  Merck

Specification Test Results

  • Assay:
    97%
  • Appearance (Colour) :
    White
  • Appearance (Form) (Powder, Crystals, Crystalline Powder, Solid, and/or Chunks):
    Conforms to Requirements
  • Infrared Spectrum:
    Conforms to Structure
  • Proton NMR Spectra:
    Conforms to Structure
  • Phosphorus NMR Spectra:
    Conforms to Structure

Specification Test Results

Frequently Bought Together