Chloride standard solution, 1,000 mg/L Cl- in water (from HCl), Titrisol®, Supelco®
Catalog # 1.09871.0001
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/l
- Matrix:H₂O
- Pk:1 Ampoul
- CAS No.:7647-01-0
- Description:Chloride standard, concentrate Titrisol®
- Seq:01
- M-Clarity quality level:MQ100
- Contents:from HCl