Order Entry
Czech Republic
ContactUsLinkComponent
Injection vials
Injection vials
Catalog # SCERG51003000F0U2
Supplier:  DWK Life Sciences
undefined
Injection vials
Catalog # SCERG51003000F0U2
Supplier:  DWK Life Sciences

Specifications

  • Cap included:
    No
  • Ø×H:
    30×100 mm
  • Capacity:
    50,00 ml
  • Seq:
    02
  • Pk:
    1.000

Specifications

About this item