
Silver 65 - 75% (Ag basis), colloidal, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-22-4
:
Formula:Ag
MDL Number:MFCD00003397
:107.87 g/mol
Boiling Pt:2212 °C (lit.)
Melting Pt:960 °C (lit.)
Density:10.49 g/cm³ (lit.)
:GHS09
Specification Test Results
- Assay (Ag basis):65 - 75%
- Form:Colloidal
Specification Test Results
Frequently Bought Together