ContactUsLinkComponent
Practice ELISA kit
Practice ELISA kit
Catalog # CAYM10009658-96
Supplier:  Cayman Chemical
CAS Number:  
undefined
Practice ELISA kit
Catalog # CAYM10009658-96
Supplier:  Cayman Chemical
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Practice ELISA kit, 96 strip wells
Specifications
About this item
A great alternative to learn Cayman’s AChE ELISA technology.
  • A practice ELISA kit for those unfamiliar with performing competitive ELISAs
  • Easy to follow instructions with helpful illustrations
This assay has been developed for researchers that do not have experience performing enzyme immunoassays (ELISA’s). It can be used as a practice tool allowing you to become comfortable with running Cayman ELISA’s. Practicing these assays can help to decrease errors when samples, time, and costs are at risk. This kit contains enough reagents to run at least four complete standard curves, including blank wells, NSB wells, and maximum binding (B0) wells. For more information regarding the development and science behind the manufacturing of ELISA’s please contact our technical service department.
Delivery: Kit contains antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover sheet, Ellman's reagent, tracer and antiserum dye.