Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Vitamin D ELISA kit

Vitamin D ELISA kit

Supplier:   Cayman Chemical
Vitamin D ELISA kit
Supplier:  Cayman Chemical
Certificates
Vitamin D ELISA kit

4 Options Available Below

Product Details & Documents

Sensitive ELISA for vitamin D quantification in plasma or serum.

  • Measure vitamin D in plasma or serum
  • Detect the metabolically stable forms 25(OH)D₃ and 25(OH)D₂
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure vitamin D levels down to 0,5 ng/ml
  • Read: Colorimetric at 405 to 420 nm

The primary synthesis of functional vitamin D begins in the skin, where a cholesterol by-product acted upon by UV light forms vitamin D₃. Vitamin D₃ is carried through the bloodstream, attached to the vitamin D binding protein, to the liver where it is converted to 25-hydroxy vitamin D₃ (25(OH)D₃). This product is then transported to the kidney where it is further converted to 1,25-dihydroxycholecalciferol (1,25(OH)₂D₃), the biologically active form. Active 1,25(OH)₂D₃ is very short-lived and is rapidly metabolized to its deactivated forms.

Vitamin D Immunoassay, accurate Vitamin D measurement in plasma or serum. Vitamin D ELISA Kit is a competitive assay that can be used for Vitamin D quantification in plasma or serum. Because of the short half-life of the biologically active 1,25(OH)D form, this Vitamin D assay primarily detects the more metabolically stable forms, 25(OH)D₃ and 25(OH)D₂.

Accurate detection of these forms requires that they be displaced from the vitamin D binding protein prior to measurement. A simple, acetone-based purification protocol is included as the initial procedure of the Vitamin D assay kit for this purpose.

Packaging: Kit contains: Antibody, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, tracer and antiserum dye.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • CAYM501050-96S
  • CAYM501050-96
  • CAYM501050-480S
  • CAYM501050-480
  • Sensitivity:
  • 0,5 ng/ml (80% B/B₀)
  • 0,5 ng/ml (80% B/B₀)
  • 0,5 ng/ml (80% B/B₀)
  • 0,5 ng/ml (80% B/B₀)
  • Detection range:
  • 0,19 - 25 ng/ml
  • 0,19 - 25 ng/ml
  • 0,19 - 25 ng/ml
  • 0,19 - 25 ng/ml
  • Mid point:
  • 1 - 4 ng/ml (50% B/B₀)
  • 1 - 4 ng/ml (50% B/B₀)
  • 1 - 4 ng/ml (50% B/B₀)
  • 1 - 4 ng/ml (50% B/B₀)
  • Developmental time:
  • 60 - 90 min
  • 60 - 90 min
  • 60 - 90 min
  • 60 - 90 min
  • Incubation time:
  • 18 hr
  • 18 hr
  • 18 hr
  • 18 hr
  • Storage temperature:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • CAYM501050-96S
    Vitamin D ELISA Kit, 96 solid wells
    Vitamin D ELISA Kit, 96 solid wells
    96 Tests
    Log in for availability
    Log in for availability
    1 * 96 Tests
    1 * 96 Tests - 19 900,00 Kč
    19 900,00 Kč
    1 * 96 Tests
       19 900,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    CAYM501050-96S
    Sensitivity
    0,5 ng/ml (80% B/B₀)
    Detection range
    0,19 - 25 ng/ml
    Mid point
    1 - 4 ng/ml (50% B/B₀)
    Developmental time
    60 - 90 min
    Incubation time
    18 hr
    Storage temperature
    –20 °C
  • CAYM501050-96
    Vitamin D ELISA Kit, 96 strip wells
    Vitamin D ELISA Kit, 96 strip wells
    96 Tests
    Log in for availability
    Log in for availability
    1 * 96 Tests
    1 * 96 Tests - 19 900,00 Kč
    19 900,00 Kč
    1 * 96 Tests
       19 900,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    CAYM501050-96
    Sensitivity
    0,5 ng/ml (80% B/B₀)
    Detection range
    0,19 - 25 ng/ml
    Mid point
    1 - 4 ng/ml (50% B/B₀)
    Developmental time
    60 - 90 min
    Incubation time
    18 hr
    Storage temperature
    –20 °C
  • CAYM501050-480S
    Vitamin D ELISA Kit, 480 solid wells
    Vitamin D ELISA Kit, 480 solid wells
    480 Tests
    Log in for availability
    Log in for availability
    1 * 480 Tests
    1 * 480 Tests - 75 590,00 Kč
    75 590,00 Kč
    1 * 480 Tests
       75 590,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    CAYM501050-480S
    Sensitivity
    0,5 ng/ml (80% B/B₀)
    Detection range
    0,19 - 25 ng/ml
    Mid point
    1 - 4 ng/ml (50% B/B₀)
    Developmental time
    60 - 90 min
    Incubation time
    18 hr
    Storage temperature
    –20 °C
  • CAYM501050-480
    Vitamin D ELISA Kit, 480 strip wells
    Vitamin D ELISA Kit, 480 strip wells
    480 Tests
    Log in for availability
    Log in for availability
    1 * 480 Tests
    1 * 480 Tests - 75 590,00 Kč
    75 590,00 Kč
    1 * 480 Tests
       75 590,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    CAYM501050-480
    Sensitivity
    0,5 ng/ml (80% B/B₀)
    Detection range
    0,19 - 25 ng/ml
    Mid point
    1 - 4 ng/ml (50% B/B₀)
    Developmental time
    60 - 90 min
    Incubation time
    18 hr
    Storage temperature
    –20 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.