Order Entry
Czech Republic
ContactUsLinkComponent
15(S)-HETE ELISA kit
15(S)-HETE ELISA kit
Supplier:  Cayman Chemical
Certificates
undefined
15(S)-HETE ELISA kit
Supplier:  Cayman Chemical

Specifications

  • Assay Type:
    Competitive
  • Target protein:
    15(S)-HETE
  • Developmental time:
    60 - 90 min
  • Mid point:
    700 - 1200 pg/ml (50% B/B₀)
  • Detection range:
    39 - 5000 pg/ml
  • Storage temperature:
    –20 °C
  • Sensitivity:
    170 pg/ml (80% B/B₀)
  • Incubation time:
    18 hr

Specifications

Related Information

Frequently Bought Together