

3-Chloroperoxybenzoic acid ≤77% (by titrimetric analysis)
Supplier: Merck
Certificates
About this item
CAS Number:937-14-4
:
Formula:C₇H₅ClO₃
MDL Number:MFCD00002127
:172.564 g/mol
Melting Pt:69…71 °C (lit.)
UN:3106
ADR:5.2,
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Proton NMR Spectrum:Conforms to Structure
- Assay:≤77%
- Impurity, 3-Chlorobenzoic Acid:≥6.0%
- Titration by Na₂S₂O₃:≤77.0%
- Water (by Karl Fischer):≥17.0%
Specification Test Results
Frequently Bought Together