ContactUsLinkComponent
BOD measurement system, BOD Direct Plus
Catalog # HACHLQV1669800001
Supplier:  Hach
undefined
BOD measurement system, BOD Direct Plus
Catalog # HACHLQV1669800001
Supplier:  Hach
Specifications
  • Range:
    0 - 40, 0 - 80, 0 - 200, 0 - 400, 0 - 800, 0 - 2000, 0 - 4000 mg/L O₂
  • Description:
    BOD Direct Plus
  • Pk:
    1 SET
Specifications
About this item
BOD Direct Plus is the next generation BOD measuring instrument that delivers accurate, automatic and direct control of wastewater samples.
  • Large user friendly graphic display
  • Delivers free individual programming of each of the samples
  • Respirometric, electronic pressure sensor measuring principle
The inductive stirring system offers maintenance and wear free operation with automatic centring of stirring rods. With the built in automatic start function there is no possibility of pressure fluctuations related to the manometric measurement.