Order Entry
Czech Republic
ContactUsLinkComponent
 
Sigma-Aldrich® HPLC injector, Rheodyne 7725/7725i
Catalog # SIAL57620-U
Supplier:  Merck
Sigma-Aldrich® HPLC injector, Rheodyne 7725/7725i
Catalog # SIAL57620-U
Supplier:  Merck

Specifications

  • Description:
    Injector, Rheodyne model 7725
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

The Rheodyne Model 7725 injector allows you to inject 1 μl to 5 ml samples with accuracy and precision.

  • Easy seal adjustment using pressure screw on front of injector
  • Wide port angle (30°), for easy access to fittings

Patented continuous flow design, flow is uninterrupted when you switch from LOAD to INJECT.

Delivery information: Injector includes a 20 μl sample loop and is supplied with a VESPEL rotor seal that can be replaced with a Tefzel rotor seal for operation at pH 0 to 14. Factory set at 5000 psi (345 bar), adjustable to 7000 psi (483 bar).

Sample loops

Product Information