Residual solvents class 2 mix C (8 components)
Catalog # REST36273
Supplier: Restek
CAS Number:
Specifications
- Concentration:In dimethyl sulfoxide
- Pk:1 Ampoul
- Description:Residual solvents class 2 mix C
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains: 2-ethoxyethanol, 800 µg/ml, ethylene glycol, 3100 µg/ml, formamide, 1100 µg/ml, N,N-dimethylacetamide, 5450 µg/ml, N,N-dimethylformamide, 4400 µg/ml, 2-methoxyethanol (methyl cellosolve), 250 µg/ml, N-methylpyrrolidone, 2650 µg/ml, sulfolane, 800 µg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.