Order Entry
Czech Republic
ContactUsLinkComponent
Phosphate buffered saline, 10X, for Western blots and IP, Sigma-aldrich®
Phosphate buffered saline, 10X, for Western blots and IP, Sigma-aldrich®
Catalog # SIALP7059-1L
Supplier:  Merck
CAS Number:  
Phosphate buffered saline, 10X, for Western blots and IP, Sigma-aldrich®
Catalog # SIALP7059-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 L
  • Description:
    10X Phosphate-Buffered Saline (PBS)
  • pH value:
    7,2 - 7,6

Specifications

About this item

Phosphate buffered saline is routinely used as a wash buffer in Western blot and immunoprecipitation procedures, as well as during sample preparation of cell lysates for use in general molecular biology applications.

Dilute 10X PBS to a 1X working solution using molecular biology grade water.

It contains 30 mM sodium phosphate, 1,5 M sodium chloride and 10,5 mM potassium phosphate.