ContactUsLinkComponent
Cleanroom wipes, Clino® one way
Cleanroom wipes, Clino® one way
Catalog # PPSA3600070
undefined
Cleanroom wipes, Clino® one way
Catalog # PPSA3600070
Specifications
  • Colour:
    White
  • L×W:
    230×230 mm
  • Pk:
    300
Specifications
About this item
Clino® one way clean surfaces in semiconductors and pharma industries.
  • More excellent intake capacity
  • High abrasion resistance
  • Low particle and fibre emission
Suitable for use in ISO class 7 and 8 (GMP grade C and D) cleanrooms.