ContactUsLinkComponent
Kontaflon 85 release agent
Kontaflon 85 release agent
  291-1245
undefined
Kontaflon 85 release agent
  291-1245
  • Capacity:
    200 ml
  • Description:
    Kontaflon 85 aerosol
  • Pk:
    1
Grease-free sliding release aerosol agent based on PTFE.
  • Constantly temperature resistant up to +260 °C
Applicable in plastics processing, vulcanizing molds and chains etc.