Zadání objednávky
Czech Republic
ContactUsLinkComponent
Synthware™ Kuderna-Danish flask
Synthware™ Kuderna-Danish flask
Catalog # SIALSYNF984250-1EA
Supplier:  Merck
Synthware™ Kuderna-Danish flask
Catalog # SIALSYNF984250-1EA
Supplier:  Merck
Specifications
  • Kapacita:
    250 ml
  • Popis:
    Flask
  • Bal.:
    1
Specifications
About this item
Flask with top joint 24/40 outer while the lower joint is a 19/22 inner.