Sand, quartz 50-70 mesh, Fluka™
:14808-60-7
:
:SiO₂
:MFCD00011232
:60.08 g/mol
:2.210 °C (1.013 hPa)
:1.710 °C
:2.650 g/cm³ (20 °C)
:Ambient
:238-878-4
:GHS08
- Particle Size:50-70 mesh
- Appearance:complying
- Granulation 0.21-0.3 mm:Min. 85 %
Frequently Bought Together